Electronic standard thermometers
Leverancier: Amarell
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
E905052 E905050
AMARE905052EA 34.8 EUR
AMARE905052 AMARE905050
Electronic standard thermometers
Thermometers Digitale thermometers
Handy and precise universal thermometer with many fields of application.
  • 'HOLD' on set-up
  • Automatic power-off after 10 minutes
  • Response time: T₉₀= 8 seconds
Probes are fast response stainless steel semiconductors (Lר: 125×3,5 mm).
Order Now
Specificaties
Environmentally Preferable