Cable ties, T, PA46
Leverancier: HellermannTyton
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HELT111-00526EA 541 EUR
HELT111-00525 HELT111-00526 HELT114-01879
Cable ties, T, PA46
Spanbanden
These inside serrated cable ties are made from polyamide 4.6 (PA46).
  • High temperature applications up to +150 (5000 h) and +195 °C (500 h)
  • Serrated inside for a strong hold onto bundles
  • Easy application either manually or with a processing tool
  • Low smoke generation in the event of a fire
  • PA46 is a moisture sensitive material.
Suitable for applications where a broader temperature range is required, such as the wind energy and rail transportation sector, electronics, defence and aerospace industry.