Avantor® Alltima, HPLC Columns, Phenyl
Leverancier: Avantor
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HICH81188EA 1160 EUR
HICH81188 HICH88087 HICH88088 HICH88092 HICH88093 96082/N 96442/N AL-5PH-300DE
Avantor® Alltima, HPLC Columns, Phenyl
Kolommen voor chromatografie
Avantor® Alltima HPLC columns are high quality phases designed for general purpose applications.
  • Base deactivated silica phases
  • Stable bonding for long column lifetime
  • Multiple selectivity options
  • USP L11
Less hydrophobic than C18 phase. Selective for aromatic compounds.