Handschoenen voor algemeen gebruik, Nitrotough™ N230B en N250B
Leverancier: Marigold (Ansell)
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
200QB0580 200QB0589 200QB0588
MARG200QB0580EA 115 EUR
MARG200QB0580 MARG200QB0589 MARG200QB0588
Handschoenen voor algemeen gebruik, Nitrotough™ N230B en N250B
Handschoenen
These cut and sewn cotton jersey gloves are available with a ¾ dipped or a full nitrile coating.
  • Interlock 100% cotton liner
  • Elastic knitwrist for snug fit
  • Very good abrasion resistance, oil repellent
  • Optimum comfort and flexibility
  • Excellent dexterity
  • Ideal alternative to PVC and light leather gloves
Typical applications include: Assembly and finishin, general site & factory maintenance, light metal fabrication and handling, metal fabrication, quality control.
Certificaten: EN 388:2016 (4111A); Cat. II