Adenosine 5'-diphosphate disodium salt (ADP disodium salt)
Leverancier: MP Biomedicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: Adenosine 5'-Diphosphate Disodium Salt, Sodium ((2R,3S,4R,5R)-5-(6-amino-9H-purin-9-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl hydrogen diphosphate
02100056.5 0210005601 0210005605
ICNA02100056.5EA 44.5 EUR
ICNA02100056.5 ICNA0210005601 ICNA0210005605
Adenosine 5'-diphosphate disodium salt (ADP disodium salt)
Adenosine 5'-diphosphate disodium salt (ADP disodium salt)
Adenosine 5′-diphosphate (ADP) is an adenine nucleotide involved in energy storage and nucleic acid metabolism via its conversion into ATP by ATP synthases. ADP affects platelet activation through its interaction with ADP receptors P2Y1, P2Y12 and P2X1. Upon its conversion to adenosine by ecto-ADPases, platelet activation is inhibited via adenosine receptors.
Storage temperature: -20 °C
Formule: C₁₀H₁₃N₅Na₂O₁₀P₂
MW: 471.167 g/mol
MDL Nummer: MFCD00066635
CAS nummer: 16178-48-6