Lecithin (from soy bean), dark yellow powder
Leverancier: MP Biomedicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
0210214780 0210214783 0210214790 0210214791
ICNA0210214780EA 34.1 EUR
ICNA0210214780 ICNA0210214783 ICNA0210214790 ICNA0210214791
Lecithin (from soy bean), dark yellow powder
Lecithin
Lecithin from Soy, Refined, is a natural mixture of phospholipids consisting of phosphatidyl choline, phosphatidyl ethanolamine, and phosphatidyl inositol. In soybean lecithin the predominant fatty acid is polyunsaturated linoleic acid, amounting to about 70% of the fatty acids present.
Lecithin is used for preparation of vesicle suspensions commonly called liposomes or as monolayers.
A major structural phospholipid in brain, comprising approx. 15% of total lipid; primarily localized to gray matter.
  • Presentation: Dark Yellow Powder
  • Storage temperature: Store at -0 °C.
Storage temperature: -20 °C
Bewaar temperatuur: Vriezer
MDL Nummer: MFCD00147406
CAS nummer: 8002-43-5
Merck Index: 12,05452
Order Now
Specification Test Results
Identity Test Passes
Source Soybean (refined oil removed)
Moisture ≤2%
Acetone Insolubles ≥95%