Multiwell cell culture plates, CELLSTAR®
Leverancier: Greiner Bio-One
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
657970_5 662970_5 677970_5
GREI657970_5EA 156 EUR
GREI657970_5 GREI662970_5 GREI677970_5
Multiwell cell culture plates, CELLSTAR®
Microplaten Celcultuurplaten
These sterile, PS multiwell cell culture plates have a cell-repellent surface to effectively inhibit cell attachment. Achieved through a chemical polymer modification, the surface does not degrade or leach under common cell culture conditions, rendering an ideal substrate for native cell culture experiments.
  • Effectively inhibits cell adhesion
  • Sterile
  • Lid with condensation rings
  • Other formats available (flasks, dishes, microplates)
Characterised by low cell attachment, the cell-repellent surface is ideal for applications such as spheroid cultures andMore Product Information