Racks for Stomacher® bags
Leverancier: Seward (FermionX)
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BA6091 BA6090 BA6096 BA6094
432-3150EA 71.8 EUR
432-3150 432-3151 432-3152 432-3153
Racks for Stomacher® bags
Rekken Zakkenrekken
Made from high grade plastic polymer.
  • Perfect for loading closure bags preenrichments into the incubator
  • Variety of capacities for each Stomacher® machine
  • Easily dismantled and cleaned with most lab disinfectants (not autoclavable)
Bag racks provide the perfect storage for Stomacher® bag sample handling, helping to provide a clean and productive working environment.