Openers voor Stomacher®-zakken
Leverancier: Seward (FermionX)
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BA6095 BA6098
431-0132EA 68.6 EUR
431-0132 432-3115
Openers voor Stomacher®-zakken
Rekken Zakkenrekken
Made from high grade plastic polymer.
  • Compatible with Stomacher® 80 bags
  • Stable on bench-top or scales
  • Easily cleaned with most lab disinfectants
These bag openers hold open bags for easy addition of sample without risk of instrument contact on the bag.