Capillary columns, HT8
Leverancier: Trajan Scientific and Medical
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
054675 054676 054677 054680 054690 054236 054682 054683
568-0206EA 840 EUR
568-0206 568-0207 568-0208 568-0210 568-0215 568-0372 568-0212 568-0002
Capillary columns, HT8
Kolommen voor chromatografie
Unique high temperature phase suited for the analysis of polychlorinated biphenyl (PCB) congeners. The unique polarity of this phase gives excellent separation of PCB congeners.
8% phenyl polycarborane-siloxane.
Maximum temperature: 360/370 °C
No equivalents.