Capillary columns, SolGel-WAX™
Leverancier: Trajan Scientific and Medical
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
054796 054791 054792 054797 054788 054789 054787 054785 054786 0547100
558-0254EA 772 EUR
558-0254 568-0221 568-0222 568-0226 568-0219 568-0220 568-0218 568-0216 568-0217 568-0425
Capillary columns, SolGel-WAX™
Kolommen voor chromatografie
These columns with polyethylene glycol (PEG) in a Sol-Gel matrix are recommended for highly active compounds.
  • Less susceptible to damage by oxygen than conventional wax phases
  • Polar phase
  • Low bleed and inert
Suitable replacement for: DB-Wax, Rtx-Wax, Stabilwax, HP20M, HP-Wax, HP-INNOWax, Supelcowax-10, AT-Wax, Nukol, CP Wax 52CB, VB-WAX, ZB-WAX.
Maximum temperature: 30 °C to 260/280 °C
Polyethylene glycol
Max. temperature: 0,1 - 1,0 µm film thickness 20 °C to 260/280 °C; >1,0 µm film thickness 20 °C to 240/260 °C