Culture media in infusion bottles
Leverancier: PHARMAMEDIA DR MULLER
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
520.S100 540.S100 550.S100
MULL520.S100EA 0 EUR
MULL520.S100 MULL540.S100 MULL550.S100
Culture media in infusion bottles
Media voor microbiologie Geprepareerde media voor microbiologie
Ready to use media for sterility test. Bottles have coloured (red or green) flip-off caps.
  • Identification and growth of aerobic micro-organisms
  • Label contains autoclaving indicator (pink to grey-brown)
  • Temperature controlled delivery
  • Label with Barcode
Certificaten: Medium according EP and USP
Verpakking: 20 bottles on a plastic tray wrapped with shrink foil