Culture media, ready to use, in plates, 90 mm with locking-lid plate
Leverancier: PHARMAMEDIA DR MULLER
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
400.0060 200.0060 220.0060
MULL400.0060EA 135 EUR
MULL400.0060 MULL200.0060 MULL220.0060
Culture media, ready to use, in plates, 90 mm with locking-lid plate
Media voor microbiologie Geprepareerde media voor microbiologie
Used in pharmaceutical industry in clean rooms and isolators for contact sampling, personnel monitoring, as well as active air monitoring. Isolation and growth of fastidious bacteria, yeasts and molds incubations in vent and closed position possible. Easy handling due to increased handling area.
  • Detection of aerobic and anaerobic micro-organisms
  • Gamma-irradiated and triple bagged
  • Locking-lid plate, made from polystyrene guarantees save transport
  • Label on side of bottom
  • High barrier foil for H₂O₂ as well as for water-vapor
Certificaten: According Ph. Eur. and Pharmacopoeia.
Leveringsinformatie: All plates are delivered in the non-locked position, and will be delivered refrigerated. Minimum order quantity 4 items.
Verpakking: 10 plates per packaging unit (6 or 10 staples)
Order Now
Specificaties
Environmentally Preferable