Electrode holders for portable meters, Orion™ Star A series
Leverancier: Thermo Orion
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
STARA-ESCD STARA-ESPH
ORIOSTARA-ESCDEA 53.1 EUR
ORIOSTARA-ESCD ORIOSTARA-ESPH
Electrode holders for portable meters, Orion™ Star A series
Elektrodehouders
These electrode holders can be used with the Star A series portable meter armor and storage sleeve to protect, transport and store sensors in the field.
  • Can be attached to either side of the portable meter armor
  • Holder for pH electrodes holds one pH electrode when placed in the pH electrode storage sleeve
  • Holder for conductivity and DO probes holds 12 mm diameter or 15 mm diameter conductivity probes and a dissolved oxygen probe when placed in its calibration sleeve
Order Now
Specificaties
Environmentally Preferable