Protective armor for portable meters, Orion™ Star A series
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ORIOSTARA-AREA 180 EUR
ORIOSTARA-AR
Protective armor for portable meters, Orion™ Star A series
Elektrodehouders
This portable meter armor adds protection around the meter screen and includes a hand or belt strap that can be converted to built-in stand, pH electrode holder, conductivity and DO probe holder and pH electrode storage sleeve.
For use with Thermo Scientific™ Orion™ Star A series portable meters (Star A320, Star A220 and Star A120 series).
Leveringsinformatie: Supplied with hand strap and built-in stand, pH electrode storage sleeve, pH electrode holder, and conductivity and dissolved oxygen sensor holder.