Microliterspuiten 5 µl
Leverancier: Trajan Scientific and Medical
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
001000 001050 001100 001400 001450 001495 001450CAL
549-0267EA 57.6 EUR
549-0267 549-0268 549-0269 549-0272 549-0273 549-0439 SGEA001450CAL
Microliterspuiten 5 µl
Spuiten Injectiespuiten voor chromatografie
These are plunger-in-barrel syringes. Each plunger is individually fitted for each microsyringe for perfect sealing.
  • Longer life
  • Superior performance and robustness
  • Reduced carryover
Plunger protection incorporates a plunger protection guide and plunger button reinforcement to minimise the risk of bent plungers.
Guided plunger microvolume syringes have an extended barrel and reinforced plunger that provides additional protection.
* SuperfleX, met flexibele zuiger. SuperfleX microvolume syringes use a super elastic alloy plunger that will not bend or kink.
Order Now
Specificaties
Environmentally Preferable