Dialysis tubing, Visking®
Leverancier: MEDICELL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
130885-25 DTV12000.01.00 DTV.12000.10
514-0009EA 117 EUR
514-0009 MEDCDTV12000.01.00 MEDCDTV.12000.10
Dialysis tubing, Visking®
Slangen Dialyseslangen
Regenerated cellulose (RC).
  • Contain small amounts of glycerol
  • Dry packaged
  • Stable at pH range 5 to 9
  • Autoclavable up to +120 °C (max. 15 min, completely submersed in dist. water)
  • No non-specific protein binding (<1 ng/g)
For laboratory analysis and ultra filtration.
Storage stability in original packaging 2 years.