Eusolex® T-AQUA
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1.05401.1000 1.05401.9030
1.05401.1000EA 0 EUR
1.05401.1000 1.05401.9030
Eusolex® T-AQUA
Eusolex®
Inorganic UV Filter
Eusolex® T-AQUA is a Titanium Dioxide based, mineral UV filter in form of a liquid, thixotropic dispersion with superior stability and free of sedimentation. This inorganic UV filter has a broad-spectrum UVA/UVB protection and is very easy to use.
INCI name: Aqua (Water), Titanium Dioxide, Alumina, Sodium Metaphosphate, Phenoxyethanol, Sodium Methylparabene
Order Now
Specificaties
Environmentally Preferable