Self-Laminating Polyester Cryo Labels B-461 Small Core 25 mm
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
217076 800537 217088 217093 217077 800536 217087 217092 217065 217097 217071 217072 217089 217086 217095
BRDY217076EA 334.94 EUR
BRDY217076 BRDY800537 BRDY217088 BRDY217093 BRDY217077 BRDY800536 BRDY217087 BRDY217092 BRDY217065 BRDY217097 BRDY217071 BRDY217072 BRDY217089 BRDY217086 BRDY217095
Self-Laminating Polyester Cryo Labels B-461 Small Core 25 mm
Etiketten
Matte, permanent self-laminating and wrap-around B-461 polyester labels for smooth surfaces. For use on conical tubes, cryo tubes and vials. Suitable for autoclaves, hot wwater baths (100 °C) and cryo or liquid nitrogen applications.
  • Die-cut labels are supplied on a roll
  • Contains some adhesive backed wire marking materials, and also labels that are pre-printed with headers or templates
  • Ultra-thin construction prevents jamming in racks and centrifuges
  • Chemical-resistant, low temperature-resistant, smear-resistant
With small core for BBP11, BBP12, THT printers.
Compliant with Directive 2011/65/EU and 2015/863
Certificaten: Compliant with Directive 2011/65/EU and 2015/863
Bestelinformatie: Supplied with the ribbon seperate, not in the same enclosure as the tape/sleeves