Cell culture flasks, cell-repellent surface, CELLSTAR®
Leverancier: Greiner Bio-One
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
690985_20 658985_15 660985_5
GREI690985_20EA 261 EUR
GREI690985_20 GREI658985_15 GREI660985_5
Cell culture flasks, cell-repellent surface, CELLSTAR®
Kolven Celcultuurflessen
These sterile, PS cell culture flasks have a cell-repellent surface to effectively inhibit cell attachment. Achieved through a chemical polymer modification, the surface does not degrade or leach under common cell culture conditions, rendering an ideal substrate for native cell culture experiments.
  • Effectively inhibits cell adhesion
  • Sterile
  • With Standard or filter screw cap
Characterised by low cell attachment, the cell-repellent surface is ideal for applications such as spheroid cultures and stem cell aggregate formation, suspension culture of semi-More Product Information