Bovine (foetal) fetuin (from serum)

Leverancier: Merck Millipore Calbiochem
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5


Calbiochem®
341506-1EA 234 EUR
341506-1
Bovine (foetal) fetuin (from serum)
Eiwitten en peptiden
Fetuin extracted from foetal bovine serum. Fetuin is an α-globulin that inhibits trypsin activity and promotes cell attachment, growth and differentiation in many different culture systems.

  • Green freeze dried solid
  • Lyophilised from 0,5% (NH₄)₂SO₄
  • Purity ≥70% by electrophoresis

Fetuin also contains both N- and O-glycosidically-linked carbohydrates. It functions as an opsonin for cationic-deactivating molecules in macrophages.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR