Premium non-stick BTO septa
Leverancier: Restek
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
27096 27084 27092 27085 27090 27091 27094 27082 27086 27087 27088 27093 27098 27099
REST27096EA 152 EUR
REST27096 REST27084 REST27092 REST27085 REST27090 REST27091 REST27094 REST27082 REST27086 REST27087 REST27088 REST27093 REST27098 REST27099
Premium non-stick BTO septa
Tussenschoten
These septa are preconditioned and ready to use. Bleed and temperature optimised; ideal for demanding GC and GC/MS applications.
  • Usable to 400 °C inlet temperature
  • New plasma coating eliminates sticking in the injection port
  • Precision molding assures consistent, accurate fit
  • CenterGuide design to minimise coring. (Not available on 9,5 mm, 10 mm and Shimadzu Plug)
  • Each batch GC-FID tested
  • 5 mm and 11 mm septa are partially predrilled
Note: For 17 mm inlets, the maximum temperature for BTO septa is 330 °C. For all injectors, minimum recommendedMore Product Information