Deep well plates, 96-well, DispoDialyzer™
Leverancier: HARVARD APPARATUS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
74-0900 74-0901 74-0903
HARV74-0900EA 4640 EUR
HARV74-0900 HARV74-0901 HARV74-0903
Deep well plates, 96-well, DispoDialyzer™
Microplaten Deepwell-microplaten
The single use 96-well DispoDialyzer™ is ideal for the simultaneous preparation of 96 samples.
  • Each well in this system has a separate membrane, thus eliminating the possibility of sample contamination and leakage
  • Very high reproducibility across different wells
  • Excellent sample recovery
  • Sealed with twelve 8-cap strips