Powder pillows, total nitrogen determination
Leverancier: Hach
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
2671846 2672046 2671946
HACH2671846EA 30.6 EUR
HACH2671846 HACH2672046 HACH2671946
Powder pillows, total nitrogen determination
Milieutestkits
For total nitrogen determination by the persulphate digestion method.