Screw thread micro-vials, with attached closures, ACCUFORM®, KIMBLE®
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SP60700-5 SP60702-10 SP60700-1 SP60700-2 SP60700-3 SP60700-10 SP60702-1
SCERSP60700-5EA 366 EUR
SCERSP60700-5 SCERSP60702-10 SCERSP60700-1 SCERSP60700-2 SCERSP60700-3 SCERSP60700-10 SCERSP60702-1
Screw thread micro-vials, with attached closures, ACCUFORM®, KIMBLE®
flesjes
Screw thread micro-vials are manufactured from 33 expansion, low extractable borosilicate glass with attached closures.
  • Tapered bases allow for maximum sample recovery
  • Internally contoured for optimal performance with limited samples
  • Graduated to ensure more accurate measurements and dispensing
Certificaten: Meets USP Type I and ASTM E438, Type I, Class A requirements.