Microwave drying oven, SAM-255
Leverancier: CEM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CEMA920085EA 0 EUR
CEMA920085
Microwave drying oven, SAM-255
Ovens Microgolfovens
SAM-255 with IntelliTemp™ infrared temperature control provides rapid drying of all types of samples. Workstation can analyse large sized samples for moisture and solids.
  • Intellitemp provides consistent drying every day, every batch
  • Works great with paint bases and colors
  • Ideal for powdered milk
  • Stores up to 20 separate methods and each method can have up to 8 heating, cooling or heat ramping stages
Time and power selections are independently programmed for each stage. The powerful, variable-speed exhaust system moves air through the cavity quickly to aid in drying and can be connected to an existing laboratory exhaust when drying organic compounds.