Screw caps, open top, without septa
Leverancier: THERMO NATIONAL SCIENTIFIC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
B7807-15 B7807-24 B7807-20
NSCAB7807-15EA 20 EUR
NSCAB7807-15 NSCAB7807-24 NSCAB7807-20
Screw caps, open top, without septa
Sluitingen Sluitingen voor chromatografievials
These PP open top screw caps can be used with sample storage screw thread vials.
  • PTFE lined solid top
  • PTFE on film/foam backing offers broad chemical resistance
  • Not autoclavable