Overhead stirrer tools, propeller, diamond, centrifugal and anchor-type impellers
Leverancier: witeg Labortechnik
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
DH.WOS501010 DH.WOS501013 DH.WOS501015 DH.WOS501017 DH.WOS501020 DH.WOS501031 DH.WOS501035 DH.WOS501111 DH.WOS501135
WITGDH.WOS501010EA 20 EUR
WITGDH.WOS501010 WITGDH.WOS501013 WITGDH.WOS501015 WITGDH.WOS501017 WITGDH.WOS501020 WITGDH.WOS501031 WITGDH.WOS501035 WITGDH.WOS501111 WITGDH.WOS501135
Overhead stirrer tools, propeller, diamond, centrifugal and anchor-type impellers
Roerders Roergereedschap
Stirring impeller for high speed-type and high torque-type overhead stirrer.
  • Made of stainless steel 304 (high-grade)
  • Resistant to acids, chemicals, corrosion and high temperature