Long flange with weld stub
Leverancier: Edwards
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
EDWAC10514316EA 112 EUR
EDWAC10514316
Long flange with weld stub
Klemmen Klemmen voor algemeen gebruik
This flange fittings made from stainless steel 316L are ideal for rugged corrosion resistance in semiconductor processing and offer excellent repeatability in high vacuum applications.
  • Quality control of elastomer specifications
  • Fit-and-forget dependability