Glass tubings, CONTURAX®
Leverancier: Schott AG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1033796 1033751 1033768 1033798 1033795
SCOR1033796EA 1300 EUR
SCOR1033796 SCOR1033751 SCOR1033768 SCOR1033798 SCOR1033795
Glass tubings, CONTURAX®
Slangen Transfer Tubing
Borosilicate glass tubing, varying types of inner and outer shapes.
  • High chemical resistance
  • High hydrolytic resistance
  • High resistance to thermal shock
For glassblown laboratory ware, pressed glass articles (e.g. balls, lenses), technical articles, and other laboratory/industrial technology.