Sampling spoons, PharmaSpoons
Leverancier: Sampling Systems
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
A504-1 A504-2 A504-3 A504-4 A504-5 A506-600 A506-1000 A506-300
SAMPA504-1EA 22.4 EUR
SAMPA504-1 SAMPA504-2 SAMPA504-3 SAMPA504-4 SAMPA504-5 SAMPA506-600 SAMPA506-1000 SAMPA506-300
Sampling spoons, PharmaSpoons
Lepels
These sampling spoons are made from stainless steel. They are ideal for transferring small amounts of powder.
  • Crevice-free design
  • Bright polished finish
  • Head is vertical and side angled (lateral) bent
Available in 316L or 304 stainless steel.
Order Now
Specificaties
Environmentally Preferable