Stirring paddles
Leverancier: Sampling Systems
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
A344-1500 A916-1000 A344-1000 A916-1500
SAMPA344-1500EA 231 EUR
SAMPA344-1500 SAMPA916-1000 SAMPA344-1000 SAMPA916-1500
Stirring paddles
Roerders Roergereedschap
These paddles are ideal for use when producing liquids and suspensions. Stirrers are made from 316 stainless steel
  • Robust construction
  • Crevice free
  • Satin finish
  • BSE/TSE free