Briggs and Salt Mixture
Leverancier: MP Biomedicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ICNA0290283410EA 737 EUR
ICNA0290283410
Briggs and Salt Mixture
Dierenbenodigdheden Voeding voor dieren
This powdered salt mixture is used as animal diet.
It contains calcium carbonate, calcium phosphate, copper sulphate, ferric citrate, magnesium sulphate, manganese sulphate, potassium chloride, potassium iodate, sodium chloride, sodium phosphate dibasic and zinc carbonate.
Opgelet: Store at room temperature.