Volumetric flasks, class A, glass, amber, DAkkS
Leverancier: GLASWARENFABRIK KARL HECHT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
92210300467 92210300546
HECH92210300467EA 351 EUR
HECH92210300467 HECH92210300546
Volumetric flasks, class A, glass, amber, DAkkS
Kolven Maatkolven
Borosilicate glass 3.3, amber, with glass stopper.
  • DAkkS calibrated
  • Calibrated to contain (TC, In)
  • White marks and inscriptions
Certificaten: DIN EN ISO 1042, DAkkS and Tolerance certified.