Ultra-low volume back pressure regulators (BPR), Upchurch Scientific®
Leverancier: UPCHURCH SCIENTIFIC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M-410 M-412 M-420
554-3073EA 748 EUR
554-3073 554-3074 UPCHM-420
Ultra-low volume back pressure regulators (BPR), Upchurch Scientific®
Ventielen Terugslagventielen
These ultra-low volume back pressure regulators (BPRs) are developed to minimise swept volume, which is especially important for multi-detector applications.
  • Completely polymeric flow path, assuring biocompatibility
With a maximum swept volume of only 6 μl, it is nearly impossible to detect these BPRs as part of the fluid pathway. To minimise the swept volume added to the flow path, it is recommended to trim the length of the attached tubing.