In-line MicroFilters, Upchurch Scientific®
Leverancier: UPCHURCH SCIENTIFIC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M-120 M-120X M-135-1 M-140 M-520 M-520-01 M-525
UPCHM-120EA 220 EUR
UPCHM-120 554-3068 UPCHM-135-1 UPCHM-140 554-3075 554-3076 UPCHM-525
In-line MicroFilters, Upchurch Scientific®
Filters Lijnfilters
These in-line MicroFilters effectively protect columns from particles originating in the mobile phase or sample, or from pump seal and sample injection valve wear.
  • 100% biocompatible PEEK polymer options available
  • Total void volume of 240 nl
  • Pressure rated to 4000 psi (276 bar)