Avantor® Hichrom, Anion Exclusion HPLC Columns
Leverancier: Avantor
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HICH269006EA 2320 EUR
HICH269006 HICH269068 38118/S
Avantor® Hichrom, Anion Exclusion HPLC Columns
Kolommen voor chromatografie
Anion exclusion columns are used for the separation of organic acids and weakly ionised anions.
  • Polymer-based for broad pH stability
  • USP L22
Mobile phases contain dilute mineral acids. Small amounts of acetonitrile are added to decrease retention of hydrophobic molecules.