PBS and TBS buffers
Leverancier: Rockland Immunochemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
MB-008 MB-011 MB-012
ROCKMB-008EA 179 EUR
ROCKMB-008 ROCKMB-011 ROCKMB-012
PBS and TBS buffers
Buffers Buffers voor elektroforese Wasbuffers
Phosphate buffered and Tris buffered saline are suitable for multiple applications including biological diluent buffer for antibodies or other biologics. Also may be used as a wash buffer for immunological assays including western blot, immunohistochemistry, immunofluorescence microscopy, and ELISA. Other applications may require detergents or other additional components.
This product is a concentrated stock solution and should be diluted appropriately with distilled, deionised water or equivalent to its final working concentration.
Store container at room temperature (18 to 26 °C) prior to opening. If desired, the solution may be stored at 4 °C or less. Some salts may precipitate out of solution at lower temperature. Allow buffer to equilibrate to room temperature (18 to 26 °C) to restore solubility of some salts.