Protein based chiral HPLC columns, CHIRALPAK® CBH
Leverancier: CHIRAL TECHNOLOGIES
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
33734 33733 33714 33713 33712 33711 33782 33783 33781 33794 33793 33792 33791 33784
554-4044EA 6730 EUR
554-4044 554-4043 554-2485 554-2484 554-2483 554-2482 554-2487 554-2488 554-2486 554-2493 554-2492 554-2491 554-2490 554-2489
Protein based chiral HPLC columns, CHIRALPAK® CBH
Kolommen voor chromatografie
These protein-based columns are suitable for separation of basic compounds. These columns function entirely in the reversed phase chromatographic mode, using buffers with low organic modifier content and at moderate pH values.
Cellobiohydrolase (CBH) is a stable enzyme, which has been immobilised onto 5 μm spherical silica particles creating the chiral stationary phase in the CHIRALPAK® CBH column. This is also a reverse phase column, used for the direct separation of enantiomers. The column is preferentially used for the separation of the enantiomers of basic drugs from many compound classes.
Narrower applicability than CHIRALPAK® AGP. Separates compounds containing one or more basic nitrogens together with one or more basic nitrogens together with one or more hydrogen accepting or hydrogen donating groups (alcohol, phenol, carbonyl, amide, ether, sulphoxide, ester, etc.).