Cation exchange chromatography columns, TSK-GEL® SP STAT
Leverancier: TOSOH Bioscience
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
0021963 0021964
TOYO0021963EA 1320 EUR
TOYO0021963 TOYO0021964
Cation exchange chromatography columns, TSK-GEL® SP STAT
Kolommen voor chromatografie
TSK-GEL® SP STAT are cross-linked hydrophilic polymer (mono-disperse particles) based columns designed to monitor a PEGylation reaction of beta-lactoglobulin. These STAT columns provide fast, high resolution separations at moderate pressures.
  • Nonporous columns
  • High efficiency separation at short analysis time
  • Used for high throughput protein separation