Reversed phase chromatography columns, RSpak DE
Leverancier: Resonac
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
F7001004 F6700150 F7001005 F7001007 F6700151
554-2918EA 1034 EUR
554-2918 554-2870 554-2919 554-2921 554-2871
Reversed phase chromatography columns, RSpak DE
Kolommen voor chromatografie
These are polymer-based columns, with similar polarity to that of ODS columns have general purpose use.
  • Usable in 100% water and buffer solutions
  • Base material is polymethacrylate