ESD-beroepsschoenen, instappers, Air Cushion, 32370
Leverancier: Abeba
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
32370/35 32370/36 32370/37 32370/38 32370/39 32370/40 32370/41 32370/42 32370/43 32370/44 32370/45 32370/46 32370/47
114-2217EA 72.7 EUR
114-2217 114-2218 114-2219 114-2220 114-2221 114-2222 114-2223 114-2224 114-2225 114-2226 114-2227 114-2228 114-2229
ESD-beroepsschoenen, instappers, Air Cushion, 32370
Schoenen en laarzen
Extremely light ESD shoe made from microfibre with a PU sole resistant to hydrocarbons.
  • Air-cushioned PU walking sole with high slip resistance
  • Instep area with elastic band
  • Sanitized® treatment reduces odour and bacterial growth
Colour: White
Kleur: Wit
Certificaten: EN 61340 (ESD); EN ISO 20347:2012; O1, SRA