Safety spectacles, Tracker
Leverancier: Bollé Safety
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
TRACPSI TRACPSF TRACWPCC5 TRACPSJ
111-0390EA 15.85 EUR
111-0390 111-0461 BOLETRACWPCC5 BOLETRACPSJ
Safety spectacles, Tracker
Brillen
These sporty design safety spectacles feature a durable PC frame, a smoke PC dual lens, straight temples with upper and lower protection and an adjustable headband.
  • Interchangeable foam reinforcement with indirect ventilation
  • Adjustable head strap and rubber arm tips provide a secure fit and greater protection
  • ALS anti-fog/anti-scratch coating on both sides of clear and smoke lenses
  • Rubberised eye seal provides exceptional protection against dust and other particles
Weight: 52 g
EN 166
Certificaten: EN 166
Leveringsinformatie: Supplied with microfibre pouch.