Test sieves, 200×50 mm
Leverancier: Retsch
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
510-5818EA 156 EUR
510-5818 510-5817 510-5816 510-5815 510-5819 510-5788 510-5821 510-5820 510-5787 510-5786 510-5785 510-5825 510-5824 510-5823 510-5789 510-5822 510-5784 510-5783 510-5782 510-5781 510-5807 510-5806 510-5805 510-5804 510-5809 510-5808 510-5810 510-5814 510-5813 510-5812 510-5811 510-5839 510-5838 510-5837 510-5843 510-5842 510-5841 510-5840 510-5847 510-5846 510-5845 510-5844 510-5829 510-5828 510-5827 510-5826 510-5799 510-5832 510-5798 510-5831 510-5797 510-5830 510-5796 510-5836 510-5835 510-5834 510-5833 510-5791 510-5790 510-5795 510-5794 510-5793 510-5792 510-5859 510-5865 510-5864 510-5863 510-5862 510-5869 510-5868 510-5867 510-5866 510-5861 510-5860 510-5849 510-5848 510-5854 510-5853 510-5852 510-5851 510-5858 510-5857 510-5856 510-5855 510-5850 510-5803 510-5802 510-5801 510-5800 510-5876 510-5875 510-5874 510-5873 510-5879 510-5878 510-5877 510-5872 510-5871 510-5870
Test sieves, 200×50 mm
Zeven Testzeven
Sieves made from 304 stainless steel are individually laser engraved with clear and precise labelling for full traceability. Available in three of the most commonly used sizes. They are fully compatible with Retsch's existing products and can be combined with other sieves without problems.
  • Sieve bodies are made in one piece and the absence of hollow spaces prevents cross-contamination
  • High corrosion resistance and easy to clean
  • Maximum stability and optimal sealing of the sieve stack, 15% less weight than traditional sieves
Special certificates available on request. Contact your local Avantor sales office for further details.
Other mesh sizes available on request.
Certificaten: ISO 3310-1:2001-09. Inspection certificate according to EN10204 3.1.B