Striped safety cleanroom tapes, SST™
Leverancier: MICRONOVA
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SST18-2BLYE SST18-2BLWH SST18-3BLYE
819-0032EA 46.4 EUR
819-0032 MICRSST18-2BLWH MICRSST18-3BLYE
Striped safety cleanroom tapes, SST™
Tape
These pressure sensitive laminated tapes are made of vinyl with rubber adhesive and are ideal for delineating and colour coding specific equipment and areas.
  • Tape is scuff resistant and holds up well to solvents, humidity and oil
  • Wound on a plastic core and cleanroom processed
Length: 16,4 m
Different colours available.
Colour: Black and yellow striped