PIG® HazMat absorbent pillows
Leverancier: NEW PIG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HR7015 PIL302 PIL306 PIL307
121-0504EA 194 EUR
121-0504 121-0540 121-0541 121-0542
PIG® HazMat absorbent pillows
Absorptiematerialen en bescherming tegen morsen
High capacity pillows pump up the absorbing power on large spills of corrosive liquids.
  • Absorbs the widest range of acids, caustics and unknown liquids, even those with high concentrations like 98% sulphuric acid and 30% sodium hydroxide
  • Chemical resistant pillow won't degrade or cause a dangerous reaction upon contact with corrosive spills
  • Pillow has large surface area, high capacity and fast wicking filler to quickly soak up liquids; great for adding absorbency during spill response
  • PP skin resists chemicals and tearing; reduces dust and holds in liquid, even when saturated
  • PP filler is highly absorbent for containing corrosive or reactive spills
The pillow absorbs and retains most acids, caustics and unknown liquids. The pink colour is easily distinguishable to assure workers that they have used the correct pillow during a spill emergency. Can be incinerated after use to reduce waste. The dispenser box makes pillows easy to access, carry and store whilst protecting contents from dirt and moisture.
Opgelet: To ensure effectiveness and your safety, we recommend that you conduct compatibility and absorption testing of your chemicals with PIG® HAZ-MAT products prior to purchase.