Cleanroom tapes, MicroSeal™ CSLB™
Leverancier: MICRONOVA
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CSLB-1WH_U CSLB-2WHIR CSLB-236WH CSLB-34WH CSLB-3WH CSLB-3WHIR CSLB-1WHIR CSLB-12WHIR CSLB-34WHIR CSLB-12WH CSLB-1WH
819-0023EA 0 EUR
819-0023 819-0025 819-0157 819-0096 819-0098 819-0105 819-0095 819-0155 819-0158 819-0154 819-0156
Cleanroom tapes, MicroSeal™ CSLB™
Tape
These cleanroom tapes are made of PE with pressure-sensitive acrylic adhesive.
  • Pre-perforated
  • Perforations at 7,6 cm (3") intervals allow an easy tear at optimum lengths
  • Low-density, low-outgassing polyethylene film will remove cleanly, leaving no adhesive residue
  • Can be written on with fibre tip and permanent markers
  • Wound on a 3" plastic core with no liner, meaning less packaging and waste to discard
  • Available from 0.5 to 3" widths; smaller widths are ideal for rodac plate sealing during environmental monitoring
  • Suitable for food processing and pharmaceutical production
  • 16,5 m roll provides approximately 42 seals
  • Irradiated tape supplied with full certification of irradiation and lot traceability
For cleanroom gowning, this conformable tape can be used to ensure an effective seal between the glove and garment. For plate sealing, bundling and labelling applications the excellent peel/reseal properties combined with the easy tear perforations make this tape the ideal choice.
Colour: White