Identification labels holder, Aesculap®
Leverancier: Aesculap
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
JG730B JG730U
AESCJG730BEA 0 EUR
AESCJG730B AESCJG730U
Identification labels holder, Aesculap®
Etikethouders
The holder for identification labels is used for mounting on trays.
  • External dimensions: 50×35×13 mm
The holder with lettering, have maximum 14 characters per line and maximum character height is 4 mm.