Dissolved oxygen sensors, InPro® 6850i
Leverancier: Mettler - Toledo
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
52206123 52206121
METN52206123EA 0 EUR
METN52206123 METN52206121
Dissolved oxygen sensors, InPro® 6850i
Elektroden Elektroden voor opgeloste zuurstof
The polarographic sensor InPro6850i is an intelligent all-rounder that uses the new 3-electrode system. A separate platinum anode increases the signal stability especially in low oxygen levels.
  • 3-electrode principle with separated platinum anode for higher signal stability
  • Permanent sensor monitoring with ISM technology reduces the risk sensor failures
  • Enhanced diagnostics allow for better maintenance planning ('as few as possible, as often as necessary')
  • FDA compliant materials and easy-to-clean high-polished surface (N5 grade) to satisfy increasing regulatory requirements
  • Reduced service time thanks to the easy-to-replace membrane body and quick-disconnect interior body
  • Durable and rugged sensor design for increased resistance to harsh environments
The integrated ISM technology with Plug and Measure capability makes installation much easier and offers new possibilities for maintenance planning thanks to automated self diagnosis and asset management.
Ideal for a wide range of applications in pharmaceutical, food and beverages and chemical industries.
Pressure resistance: 0,2 to 6 bar
Certificaten: ATEX: EEx II 1/2G IIC T6/T5/T4/T3