Cation exchange chromatography columns, TSK-GEL® CM-3SW
Leverancier: Tosoh Bioscience
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
TSK-Gel®
TOYO0007650EA 892 EUR
TOYO0007650 TOYO0007167 TOYO0007162
Cation exchange chromatography columns, TSK-GEL® CM-3SW
Kolommen voor chromatografie
TSK-GEL® CM-3SW are silica based stainless steel columns packed with spherical silica beads which are modified with carboxymethyl (CM) ligands to provide a weak cation exchange column. Designed for the separation and analysis of proteins, peptides, and other biologically active molecules.